Anti-Growth Hormone Rabbit Polyclonal Antibody
BSBTRP1007
- Antibody type:Primary
- Antigen name:Growth hormone 1
- Antigen symbol:GH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Neutralisation:Yes
- Form:Lyophilized
- Antigen synonyms:GH-N|GH 1|GH|GHN|Growth Hormone 1|hGH-N|GH-1|IGHD1B
- Amino acid number:27 - 216
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:E. coli-derived rat GH recombinant protein (Position: F27-F216)
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Somatotropin(GH1) detection. Tested with WB, IHC-P, ELISA, Neu, IP in Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Growth Hormone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat