Anti-BCL-2 Rabbit Polyclonal Antibody
Catalog # BSBTRP1003
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:B-cell lymphoma 2
- Antigen symbol:BCL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Neutralisation:Yes
- Form:Lyophilized
- Antigen synonyms:PPP1R50|Bcl-2
- Amino acid number:1 - 211
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:E. coli-derived human BCL-2 recombinant protein (Position: M1-D211)
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Apoptosis regulator Bcl-2(BCL2) detection. Tested with WB, ELISA, Neu, IP in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: BCL-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human