Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-E2F4 Rabbit Polyclonal Antibody
  BSBTPA2156
undefined
Anti-E2F4 Rabbit Polyclonal Antibody
  BSBTPA2156

 

  • Antibody type:
    Primary
  • Antigen symbol:
    E2F4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    E2F-4
  • Amino acid number:
    228 - 243
  • Sequence:
    LPKPALAQSQEASRPN
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human E2F4 (228-243aa LPKPALAQSQEASRPN), different from the related mouse and rat sequences by four amino acids
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Transcription factor E2F4(E2F4) detection. Tested with WB in Human, Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: E2F4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat