Anti-Aromatase Rabbit Polyclonal Antibody
Catalog # BSBTPA2107
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450 Family 19 Subfamily A Member 1
- Antigen symbol:CYP19A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:ARO, ARO1, CPV1, CYAR, CYP19, CYPXIX, P-450AROM, Aromatase
- Amino acid number:455 to 476
- Sequence:LKRFHVKTLQKRCIENMPKNND
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of rat Aromatase (455-476aa LKRFHVKTLQKRCIENMPKNND).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Aromatase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Rat