Anti-ADAMTS1 Rabbit Polyclonal Antibody
BSBTPA2071
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase with Thrombospondin Type 1 Motif, 1
- Antigen symbol:ADAMTS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:ADAMTS|ADAM-TS1|METH1|ADAMTS-1|C3-C5|METH-1
- Amino acid number:956 - 968
- Sequence:KHYIDFCTLTQCS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of mouse ADAMTS1 (956-968aa KHYIDFCTLTQCS), different from the related rat sequence by one amino acid, and from the related human sequence by four amino acids
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for A disintegrin and metalloproteinase with thrombospondin motifs 1(ADAMTS1) detection. Tested with WB in Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: ADAMTS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat