Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PKM2 Rabbit Polyclonal Antibody
Anti-PKM2 Rabbit Polyclonal Antibody
  BSBTPA2046
undefined
Anti-PKM2 Rabbit Polyclonal Antibody
  BSBTPA2046

 

  • Antibody type:
    Primary
  • Antigen name:
    Pyruvate Kinase, muscle
  • Antigen symbol:
    PKM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    THBP1|PKM2|TCB|CTHBP|HEL-S-30|PK3|OIP3
  • Amino acid number:
    77 - 91
  • Sequence:
    SHGTHEYHAETIKNV
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human PKM2 (77-91aa SHGTHEYHAETIKNV), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Pyruvate kinase isozymes M1/M2 (PKM) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: PKM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat