Anti-Stanniocalcin 1 Rabbit Polyclonal Antibody
BSBTPA1997
- Antibody type:Primary
- Antigen name:Stanniocalcin 1
- Antigen symbol:STC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:STC
- Amino acid number:150 to 167
- Sequence:QLPNHFSNRYYNRLVRSL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Stanniocalcin 1 (150-167aa QLPNHFSNRYYNRLVRSL), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: Stanniocalcin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat