Anti-p107 Rabbit Polyclonal Antibody
Catalog # BSBTPA1894
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Retinoblastoma-like 1
- Antigen symbol:p107
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:RBL1_HUMAN.|PRB1|RBL1phospho S975|CP107|Retinoblastoma like protein 1|RBL1|Cellular protein 107
- Amino acid number:1048 - 1068
- Sequence:QENDDVLLKRLQDVVSERANH
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human p107 (1048-1068aa QENDDVLLKRLQDVVSERANH), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Retinoblastoma-like protein 1 (RBL1) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: p107
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat