Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CD1a Rabbit Polyclonal Antibody
  BSBTPA1875
undefined
Anti-CD1a Rabbit Polyclonal Antibody
  BSBTPA1875

 

  • Antibody type:
    Primary
  • Antigen name:
    CD1a molecule
  • Antigen symbol:
    CD1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    FCB6|cortical thymocyte antigen CD1A|T-cell surface antigen T6/Leu-6|cluster of differentiation 1 A|T6|CD1|R4|a polypeptide|T-cell surface glycoprotein CD1a|CD1A antigen|HTA1
  • Amino acid number:
    158 - 173
  • Sequence:
    AKHFCKVLNQNQHEND
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human CD1a (158-173aa AKHFCKVLNQNQHEND).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for T-cell surface glycoprotein CD1a (CD1A) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CD1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human