Anti-CD163 Rabbit Polyclonal Antibody
Catalog # BSBTPA1874-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:CD163 molecule
- Antigen symbol:CD163
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:CD163 antigen|scavenger receptor cysteine-rich type 1 protein M130|hemoglobin scavenger receptor|macrophage-associated antigen
- Amino acid number:1056 - 1165
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:E. coli-derived human CD163 recombinant protein (Position: F1056-L1165).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Scavenger receptor cysteine-rich type 1 protein M130 (CD163) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: CD163
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human