Anti-IRF3 Rabbit Polyclonal Antibody
Catalog # BSBTPA1819
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:IRF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:409 - 427
- Sequence:KAYLQDLVEGMDFQGPGES
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human IRF3 (409-427aa KAYLQDLVEGMDFQGPGES), different from the related mouse and rat sequences by five amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Interferon regulatory factor 3 (IRF3) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: IRF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human