Anti-Daxx Rabbit Polyclonal Antibody
BSBTPA1809
- Antibody type:Primary
- Antigen name:Death-associated protein 6
- Antigen symbol:DAXX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:Fas death domain-associated protein|MGC126246|BING2|EAP1|ETS1-associated protein 1|DAP6|MGC126245|Death domain-associated protein 6|DADB-159G18.9
- Amino acid number:325 - 341
- Sequence:QERRHLDLIYNFGCHLT
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Daxx (325-341aa QERRHLDLIYNFGCHLT), identical to the related mouse and rat sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Death domain-associated protein 6 (DAXX) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Daxx
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat