Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Aryl hydrocarbon Receptor Rabbit Polyclonal Antibody
Anti-Aryl hydrocarbon Receptor Rabbit Polyclonal Antibody
Katalogové číslo BSBTPA1782-1
Dodavatel:  Boster Bio
undefined
Anti-Aryl hydrocarbon Receptor Rabbit Polyclonal Antibody
Katalogové číslo BSBTPA1782-1
Dodavatel:  Boster Bio

Specifikace

  • Antibody type:
    Primary
  • Antigen name:
    Aryl Hydrocarbon Receptor
  • Antigen symbol:
    AHR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    BHLHE76|AHR|Ah receptor|AhR|Class E basic helix-loop-helix protein 76|bHLHe76
  • Amino acid number:
    832 - 848
  • Sequence:
    HPSEARPFPDLTSSGFL
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Aryl hydrocarbon Receptor (832-848aa HPSEARPFPDLTSSGFL).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifikace

O tomto produktu

Rabbit IgG polyclonal antibody for Aryl hydrocarbon receptor (AHR) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Aryl hydrocarbon Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human