Anti-MCM5 Rabbit Polyclonal Antibody
Catalog # BSBTPA1653
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Minichromosome Maintenance Complex Component 5
- Antigen symbol:MCM5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:P1-CDC46|MGC5315|CDC46
- Amino acid number:720 - 734
- Sequence:EIQHRMQRKVLYRLK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human MCM5 (720-734aa EIQHRMQRKVLYRLK), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for DNA replication licensing factor MCM5(MCM5) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: MCM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat