Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Caspase 9 Rabbit Polyclonal Antibody
  BSBTPA1595
undefined
Anti-Caspase 9 Rabbit Polyclonal Antibody
  BSBTPA1595

 

  • Antibody type:
    Primary
  • Antigen name:
    Caspase 9
  • Antigen symbol:
    CASP9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
  • Amino acid number:
    183 - 198
  • Sequence:
    SNIDCEKLRRRFSSLH
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human Caspase 9 (183-198aa SNIDCEKLRRRFSSLH).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Caspase-9(CASP9) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Caspase 9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human