Anti-Eph Receptor B3 Rabbit Polyclonal Antibody
Catalog # BSBTPA1569
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:EPH Receptor B3
- Antigen symbol:EPHB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:EPHB3_HUMAN|Ephrin type-B receptor 3|Ephrin type B receptor 3|Cek10|Sek4|EPH Like Tyrosine Kinase 2|Embryonic kinase 2|EPH-like kinase 2|Tyrosine protein kinase TYRO6|EK2|Tyrosine-protein kinase TYRO6.|Ephrin receptor EphB3|Tyrosine protein kinase receptor HEK2|Human Embryo Kinase 2|TYRO6|Mdk5|hEK2|ETK2|ephb3
- Amino acid number:982 - 998
- Sequence:SIQDMRLQMNQTLPVQV
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Eph receptor B3 (982-998aa SIQDMRLQMNQTLPVQV), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Ephrin type-B receptor 3(EPHB3) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Eph Receptor B3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat