Anti-CD89 Rabbit Polyclonal Antibody
BSBTPA1549
- Antibody type:Primary
- Antigen name:Fc fragment of IgA receptor
- Antigen symbol:FCAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:FCAR_HUMAN|Immunoglobulin alpha Fc receptor.|Fc alpha receptor|CD89|IgA Fc receptor|FCAR
- Amino acid number:84 - 101
- Sequence:EFVIDHMDANKAGRYQCQ
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle of human CD89 (84-101aa EFVIDHMDANKAGRYQCQ).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Immunoglobulin alpha Fc receptor(FCAR) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: CD89
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human