Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) receptor 4
- Antigen symbol:CCR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:ChemR13|CC-CKR-4|CKR4|HGCN:14099|K5-5|CMKBR4|CD194
- Amino acid number:346 to 360
- UniProtKB:P51680
- Sequence:SYTQSTVDHDFRDAL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of mouse CCR4 (346-360aa, SYTQSTVDHDFRDAL), identical to the related rat sequence.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for C-C chemokine receptor type 4(CCR4) detection. Tested with WB in Mouse;Rat.
CCR4,C-C chemokine receptor type 4 is a protein that in humans is encoded by the CCR4 gene. CCR4 has also recently been designated CD194(cluster of differentiation 194). The protein encoded by this gene belongs to the G protein-coupled receptor family. It is a receptor for the following CC chemokines. The CCR4 gene is mapped to chromosome 3. Chemokines are a group of small structurally related proteins that regulate cell trafficking of various types of leukocytes. The chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: CCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat