Anti-CD90/Thy1 Rabbit Polyclonal Antibody
BSBTPA1515-1
- Antibody type:Primary
- Antigen name:Thy1 Cell Surface Antigen
- Antigen symbol:THY1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Thy-1 T-cell antigen|Thy-1 cell surface antigen|Thy-1 membrane glycoprotein|THY1|FLJ33325
- Amino acid number:20 - 39
- Sequence:QKVTSLTACLVDQSLRLDCR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human CD90 (20-39aa QKVTSLTACLVDQSLRLDCR), different from the related mouse sequence by two amino acids, and from the related rat sequence by four amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Thy-1 membrane glycoprotein(THY1) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: CD90/Thy1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat