Anti-MEKK2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1499
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase kinase 2
- Antigen symbol:MAP3K2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:MEKK2B|MEK kinase 2|MAP/ERK kinase kinase 2|MEKK2|MEKK 2|MAPK/ERK kinase kinase 2
- Amino acid number:606 - 619
- Sequence:SADELLRHMFVHYH
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human MEKK2 (606-619aa SADELLRHMFVHYH), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Mitogen-activated protein kinase kinase kinase 2(MAP3K2) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: MEKK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat