Anti-IL23 P19 Rabbit Polyclonal Antibody
Catalog # BSBTPA1470
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 23 Alpha
- Antigen symbol:IL23A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:P19|SGRF|IL23P19|IL-23|IL-23A
- Amino acid number:29 - 48
- Sequence:DWAQCQQLSRNLCMLAWNAH
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of mouse IL23 P19 (29-48aa DWAQCQQLSRNLCMLAWNAH), different from the related rat sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Interleukin-23 subunit alpha(IL23A) detection. Tested with WB in Mouse.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: IL23 P19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse