Anti-IL12 p40 Rabbit Polyclonal Antibody
Catalog # BSBTPA1468
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 12 Beta
- Antigen symbol:IL12B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:23 - 40
- Sequence:IWELKKDVYVVELDWYPD
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human IL12 p40 (23-40aa IWELKKDVYVVELDWYPD).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Interleukin-12 subunit beta(IL12B) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: IL12 p40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human