Anti-ADAMTS2 Rabbit Polyclonal Antibody
BSBTPA1461
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase With Thrombospondin Type 1 Motif, 2
- Antigen symbol:ADAMTS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:ADAMTS-2|NPI|PNPI|PCINP|PCPNI|ADAM-TS2|ADAMTS-3|PC I-NP|PCI-NP
- Amino acid number:1189 to 1204
- Sequence:KTRNQRIQELIDEMRK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human ADAMTS2 (1189-1204aa KTRNQRIQELIDEMRK), different from the related rat and mouse sequences by three amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: ADAMTS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat