Anti-DDT Rabbit Polyclonal Antibody
Catalog # BSBTPA1449
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:D-dopachrome tautomerase
- Antigen symbol:DDT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:DDCT
- Amino acid number:99 - 118
- Sequence:RFFPLESWQIGKIGTVMTFL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human DDT (99-118aa RFFPLESWQIGKIGTVMTFL), different from the related mouse and rat sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for D-dopachrome decarboxylase(DDT) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: DDT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat