Anti-IL-4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1424
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 4
- Antigen symbol:IL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:BCGF-1|B-cell stimulatory factor 1|IL-4|Interleukin 4|BCGF1|BSF1|BSF-1|Interleukin-4
- Amino acid number:72 - 98
- Sequence:AATVLRQFYSHHEKDTR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human IL4 (72-98aa AATVLRQFYSHHEKDTR).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Interleukin-4(IL4) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: IL-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human