Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MEK3 Rabbit Polyclonal Antibody
Anti-MEK3 Rabbit Polyclonal Antibody
Catalog # BSBTPA1377
Supplier:  Boster Bio
undefined
Anti-MEK3 Rabbit Polyclonal Antibody
Catalog # BSBTPA1377
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dual specificity mitogen-activated protein kinase kinase 3
  • Antigen symbol:
    MAP2K3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    SAPKK-2|MEK 3|MAP kinase kinase 3|MEK3|SAPK kinase 2|SAPKK2|PRKMK3|SKK2|MKK3|MAPKK 3|MAP2K3|Stress-activated protein kinase kinase 2|MAPK/ERK kinase 3|Dual specificity mitogen-activated protein kinase kinase 3
  • Amino acid number:
    320 - 334
  • Sequence:
    MEHPFFTLHKTKKTD
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human MEK3 (320-334aa MEHPFFTLHKTKKTD), identical to the related mouse and rat sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Dual specificity mitogen-activated protein kinase kinase 3(MAP2K3) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: MEK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat