Anti-CD80 Rabbit Polyclonal Antibody
Catalog # BSBTPA1365-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:CD80 molecule
- Antigen symbol:CD80
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:TSA1|B71|Ly-53|MIC17|Cluster of Differentiation 80|Cd28l|Ly53
- Amino acid number:227 - 245
- Sequence:DAHVSQNFTWEKPPEDPPD
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of rat CD80 (227-245aa DAHVSQNFTWEKPPEDPPD), different from the related mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for T-lymphocyte activation antigen CD80(CD80) detection. Tested with WB in Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: CD80
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Rat