Anti-P cadherin Rabbit Polyclonal Antibody
Catalog # BSBTPA1363
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:P Cadherin
- Antigen symbol:P cadherin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:612 - 626
- Sequence:QDTYDVHLSLSDHGN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human P cadherin (612-626aa QDTYDVHLSLSDHGN), different from the mouse and rat sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Cadherin-3(CDH3) detection. Tested with WB, IHC-P, ICC in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: P cadherin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human