Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Superoxide Dismutase 1 Rabbit Polyclonal Antibody
Anti-Superoxide Dismutase 1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1345
Supplier:  Boster Bio
undefined
Anti-Superoxide Dismutase 1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1345
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Superoxide Dismutase 1
  • Antigen symbol:
    SOD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    Cu/Zn SOD|ALS|SOD|ALS1|Cu/Zn superoxide dismutase|Amyotrophic lateral sclerosis 1 adult|Homodimer|Amyotrophic lateral sclerosis 1|ALS 1
  • Amino acid number:
    113 - 129
  • Sequence:
    IIGRTLVVHEKADDLGK
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of Human Superoxide Dismutase 1 (113-129aa IIGRTLVVHEKADDLGK), different from the mouse sequence by two amino acids
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Superoxide dismutase [Cu-Zn](SOD1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Superoxide Dismutase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat