Anti-nNOS (neuronal) Rabbit Polyclonal Antibody
BSBTPA1329
- Antibody type:Primary
- Antigen name:Nitric Oxide Synthase 1
- Antigen symbol:NNOS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:bNOS1|Neuronal Nitric Oxide Synthase|NOS Type 1|Brain NOS
- Amino acid number:1418 - 1434
- Sequence:IAFIEESKKDTDEVFSS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human nNOS (neuronal) (1418-1434aa IAFIEESKKDTDEVFSS), identical to the related mouse and rat sequences
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Nitric oxide synthase, brain (NOS1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: nNOS (neuronal)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat