Anti-Notch1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1215
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Notch 1
- Antigen symbol:NOTCH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:transmembrane receptor Notch1|lin-12,Motch A|RP23-306D20.12-001|N1|9930111A19Rik|neurogenic locus notch homolog protein 1|major type A protein|Tan1|p300|Mis6|Notch gene homolog 1|mT14
- Amino acid number:2487 - 2506
- Sequence:QHSYSSPVDNTPSHQLQVPE
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Notch1 (2487-2506aa QHSYSSPVDNTPSHQLQVPE)
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Neurogenic locus notch homolog protein 1 (NOTCH1) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Notch1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human