Anti-COX2/Cyclooxygenase 2 Rabbit Polyclonal Antibody
BSBTPA1211
- Antibody type:Primary
- Antigen name:Cyclooxygenase 2
- Antigen symbol:COX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:26 - 41
- Sequence:CQNRGVCMSVGFDQYK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human COX2 (26-41aa CQNRGVCMSVGFDQYK), different from the related rat sequence by two amino acids
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Prostaglandin G/H synthase 2 (PTGS2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: COX2/Cyclooxygenase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat