Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PP2A Rabbit Polyclonal Antibody
Anti-PP2A Rabbit Polyclonal Antibody
  BSBTPA1068
undefined
Anti-PP2A Rabbit Polyclonal Antibody
  BSBTPA1068

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein Phosphatase 2, Catalytic Subunit, alpha isozyme
  • Antigen symbol:
    PP2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Amino acid number:
    6 - 20
  • Sequence:
    FTKELDQWIEQLNEC
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human PP2A (6-20aa FTKELDQWIEQLNEC), identical to the related rat and mouse sequences
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform (PPP2CA) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: PP2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat