Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-Myeloperoxidase Rabbit Polyclonal Antibody
Anti-Myeloperoxidase Rabbit Polyclonal Antibody
  BSBTPA1054
undefined
Anti-Myeloperoxidase Rabbit Polyclonal Antibody
  BSBTPA1054

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Myeloperoxidase
  • Symbol antigenu:
    MPO
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    C-Terminal
  • Křížová adsorbce:
    No
  • Formulář:
    Lyophilised
  • Počet aminokyselin:
    714 - 728
  • Sekvence:
    KNNIFMSNSYPRDFV
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    Ice
  • Imunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Myeloperoxidase (714-728aa KNNIFMSNSYPRDFV), different from the related mouse and rat sequences by one amino acid
  • Čištění:
    purified by immunogen affinity purification
  • Rozměry protilátky:
    100 µg
  • Bal.:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Myeloperoxidase (MPO) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Myeloperoxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat