Order Entry
Czech Republic
ContactUsLinkComponent
Anti-alpha-Adaptin Mouse Monoclonal Antibody [clone: Ada-1]
Anti-alpha-Adaptin Mouse Monoclonal Antibody [clone: Ada-1]
  BSBTMA1105
Anti-alpha-Adaptin Mouse Monoclonal Antibody [clone: Ada-1]
  BSBTMA1105

 

  • Antibody type:
    Primary
  • Antigen name:
    alpha Adaptin
  • Clonality:
    Monoclonal
  • Clone:
    Ada-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    AP-2 adaptor polypeptides from bovine brain.
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: alpha-Adaptin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat