Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Troponin T Mouse Monoclonal Antibody [clone: TT-98]
Anti-Troponin T Mouse Monoclonal Antibody [clone: TT-98]
Catalog # BSBTMA1098
Supplier:  Boster Bio
undefined
Anti-Troponin T Mouse Monoclonal Antibody [clone: TT-98]
Catalog # BSBTMA1098
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Troponin T
  • Antigen symbol:
    TNNT2
  • Clonality:
    Monoclonal
  • Clone:
    TT-98
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Rabbit,
    Chicken,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Antigen synonyms:
    TnTC|CMPD2|LVNC6|CMH2|cTnT|CMD1D|RCM3
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Troponin T from rabbit skeletal muscle
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Mouse IgG monoclonal antibody for Troponin T, troponin T type 1 (skeletal, slow); troponin T type 2 (cardiac); troponin T type 3 (skeletal, fast) (TNNT1; TNNT2; TNNT3) detection. Tested with WB, IHC-P, IHC-F in Human;mouse;rat;rabbit;chicken. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Troponin T
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat, Rabbit, Chicken