Anti-N-Cadherin Mouse Monoclonal Antibody [clone: NC-17]
Catalog # BSBTMA1067
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cadherin 2
- Antigen symbol:CDH2
- Clonality:Monoclonal
- Clone:NC-17
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Chicken,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Antigen synonyms:CDHN|Neural-Cadherin|CDw325|N-Cadherin|CDH2|NCAD
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Affinity purified chicken heart A-CAM
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for N-Cadherin, cadherin 2, type 1, N-cadherin (neuronal) (CDH2) detection. Tested with WB, IHC-F in Human;mouse;rat;rabbit;chicken. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: N-Cadherin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit, Chicken