Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Myosin (Skeletal, Fast) Mouse Monoclonal Antibody [clone: MY-38]
Anti-Myosin (Skeletal, Fast) Mouse Monoclonal Antibody [clone: MY-38]
Catalog # BSBTMA1063
Supplier:  Boster Bio
undefined
Anti-Myosin (Skeletal, Fast) Mouse Monoclonal Antibody [clone: MY-38]
Catalog # BSBTMA1063
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myosin (Skeletal, Fast)
  • Antigen symbol:
    Myosin (Skeletal, Fast)
  • Clonality:
    Monoclonal
  • Clone:
    MY-38
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Rabbit,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilised
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Rabbit muscle myosin
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Mouse IgG monoclonal antibody for Myosin (Skeletal, Fast), myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 2, skeletal muscle, adult (MYH1; MYH2) detection. Tested with WB, IHC-P in Human;mouse;rat;rabbit. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Myosin (Skeletal, Fast)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit