Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Proteasome 20S beta 6 Rabbit Monoclonal Antibody [clone: EPR9685]
Anti-Proteasome 20S beta 6 Rabbit Monoclonal Antibody [clone: EPR9685]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-Proteasome 20S beta 6 Rabbit Monoclonal Antibody [clone: EPR9685]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB150366-1ML
  • ABCAAB150366-100
  • Antigen name:
  • Proteasome 20S beta 6
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Host:
  • Rabbit
  • Western blot:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together