- Antibody type:Primary
- Antigen name:solute carrier family 38, member 1
- Antigen symbol:Solute carrier family 38, member 1
- Clonality:Monoclonal
- Clone:N104-32
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Seq:1286
- Form:Liquid
- Antigen synonyms:Solute carrier family 38 member 1, S|Amino acid transporter A1|N-system amino acid transporter 2|Sodium-coupled neutral amino acid transporter 1
- Storage buffer:Storage buffer
Preservative: 0,09% Sodium azide
Constituents: 50% Glycerol (glycerin, glycerine), 49% PBS - Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:WB
ImmunoChemistry - Size:100 µg
- Pack type:Vial
- Pk:100 µG
Mouse monoclonal [N104/32] to SLC38A1/NAT2.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SLC38A1/NAT2
Clonality: Monoclonal
Clone: N104/32
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat, Human