Anti-EGFR Mouse Monoclonal Antibody [clone: EGFR1] (PE (Phycoerythrin))
Catalog # ABCAAB130738-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Epidermal growth factor receptor
- Antigen symbol:EGFR
- Clonality:Monoclonal
- Clone:EGFR1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pack type:Vial
- Pk:100 Tests
Specifications
About this item
PE Mouse monoclonal [EGFR1] to EGFR.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: EGFR1
Conjugation: PE (Phycoerythrin)
Epitope: Within amino acids 6-273
Host: Mouse
Isotype: IgG2b
Reactivity: Horse;Human