Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KDM1/LSD1 Rabbit Monoclonal Antibody [clone: EPR6825]
Anti-KDM1/LSD1 Rabbit Monoclonal Antibody [clone: EPR6825]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-KDM1/LSD1 Rabbit Monoclonal Antibody [clone: EPR6825]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB129195-20UL
  • ABCAAB129195-1ML
  • ABCAAB129195-100
  • Antigen symbol:
  • KDM1A
  • Antigen name:
  • lysine (K)-specific demethylase 1A
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR6825
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together