Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MAD2L1 Mouse Monoclonal Antibody [clone: 4D2]
Anti-MAD2L1 Mouse Monoclonal Antibody [clone: 4D2]
Catalog # ABCAAB114036-100
Supplier:  Abcam
Anti-MAD2L1 Mouse Monoclonal Antibody [clone: 4D2]
Catalog # ABCAAB114036-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAD2 mitotic arrest deficient-like 1 (yeast)
  • Antigen symbol:
    MAD2L1
  • Clonality:
    Monoclonal
  • Clone:
    4D2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MAD2L1 (MAD2 mitotic arrest deficient-like 1 (yeast))
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human