Anti-HMGCL Mouse Monoclonal Antibody [clone: 9F5AB9]
ABCAAB110295-100
: Abcam
- Druh protilátky:Primární
- Název antigenu:3-hydroxymethyl-3-methylglutaryl-CoA lyase
- Symbol antigenu:HMGCL
- Klonovalita:Monoclonal
- Klon:9F5AB9
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Imunoprecipitace:Yes
- Izotyp:IgG2b
- Sekvence:1286
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Typ balení:Vial
- Bal.:100 µG
Mouse monoclonal [9F5AB9] to HMGCL.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: HMGCL
Clonality: Monoclonal
Clone: 9F5AB9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human