Anti-Proteasome 20S alpha 2/HC3 Rabbit Monoclonal Antibody [clone: EPR5454]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR5454] to Proteasome 20S alpha 2/HC3.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Proteasome 20S alpha 2/HC3
Clonality: Monoclonal
Clone: EPR5454
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCAAB109502-1ML
- ABCAAB109502-100
- Antigen symbol:
- Proteasome 20S alpha 2
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- EPR5454
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- Flow cytometry:
- Yes
- Storage temperature:
- Cool
- Shipping temperature:
- Ice
Specifications
Frequently Bought Together