Anti-Complement factor B Mouse Monoclonal Antibody [clone: M13/12]
Product Details & Documents
Mouse Monoclonal CFAB antibody. Suitable for IP, WB and reacts with Human samples. Cited in 2 publications. Immunogen corresponding to Full Length Protein corresponding to Human CFB.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Complement factor B
Clonality: Monoclonal
Clone: M13/12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Seq:1202
- Size:50 µg
- Pack type:Vial
- Western blot:Yes
- Clonality:Monoclonal
- ImmunoPrecipitation:Yes
- Purification:Protein G Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG1
- Storage temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Host:Mouse
- Concentration:0,1 mg/ml
- Molecular weight:85533 Da
- Shipping temperature:4 °C
- Antigen name:BF
- Cat. No.:ABCAAB106139-50
- Reactivity:Human
- Antigen synonyms:BFD|CFB|C3/C5 convertase|Glycine-rich beta glycoprotein|PBF2|Properdin factor B|GBG
- Immunogen:Full Length Protein corresponding to Human CFB. The exact immunogen used to generate this antibody is proprietary information.
- Conjugation:Unconjugated
- Tested applications:IP
- Antigen symbol:Complement factor B
- Storage buffer:PBS, BSA
- Clone:M13/12



