Anti-MUC1 Mouse Monoclonal Antibody [clone: BC-2]
Catalog # ABCAAB89492-500
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:mucin 1, cell surface associated
- Antigen symbol:MUC1
- Clonality:Monoclonal
- Clone:BC-2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µl
- Pack type:Vial
- Pk:500 µl
Specifications
About this item
Mouse monoclonal [BC-2] to MUC1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: BC-2
Conjugation:
Epitope: Reacts with a 5 amino acid epitope of the MUC1 core protein which is less affective for glycosilation than the epitopes detected by other MUC1 antibodies
Host: Mouse
Isotype: IgG1
Reactivity: Human