Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cdc4 / Fbw7 / hSel 10 Mouse Monoclonal Antibody [clone: FBOX 3a9/1]
Anti-Cdc4 / Fbw7 / hSel 10 Mouse Monoclonal Antibody [clone: FBOX 3a9/1]
Catalog # ABCAAB74054-100
Supplier:  Abcam
Anti-Cdc4 / Fbw7 / hSel 10 Mouse Monoclonal Antibody [clone: FBOX 3a9/1]
Catalog # ABCAAB74054-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CDC4
  • Clonality:
    Monoclonal
  • Clone:
    FBOX 3a9/1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    Fbw7 / hSel 10
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cdc4 / Fbw7 / hSel 10
Clonality: monoclonal
Clone: FBOX 3a9/1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse