Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HIV1 Nef Mouse Monoclonal Antibody [clone: JR6]
Anti-HIV1 Nef Mouse Monoclonal Antibody [clone: JR6]
Catalog # ABCAAB42358-100
Supplier:  Abcam
undefined
Anti-HIV1 Nef Mouse Monoclonal Antibody [clone: JR6]
Catalog # ABCAAB42358-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HIV1 Nef
  • Clonality:
    Monoclonal
  • Clone:
    JR6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [JR6] to HIV1 Nef.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HIV1 Nef
Clonality: Monoclonal
Clone: JR6
Conjugation:
Epitope: Mapped to amino acids 195-205 (ARELHPEYYKD - B subtype consensus)
Host: Mouse
Isotype: IgG1
Reactivity: HIV