Order Entry
Czech Republic
ContactUsLinkComponent
Anti-beta subunit Cholera Toxin Mouse Monoclonal Antibody [clone: 2/63]
Anti-beta subunit Cholera Toxin Mouse Monoclonal Antibody [clone: 2/63]
Catalog # ABCAAB35988-200
Supplier:  Abcam
undefined
Anti-beta subunit Cholera Toxin Mouse Monoclonal Antibody [clone: 2/63]
Catalog # ABCAAB35988-200
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta subunit Cholera Toxin
  • Clonality:
    Monoclonal
  • Clone:
    2/63
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µg
  • Pack type:
    Vial
  • Pk:
    200 µG

Specifications

About this item

Mouse monoclonal [2/63] to beta subunit Cholera Toxin.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: beta subunit Cholera Toxin
Clonality: Monoclonal
Clone: 2/63
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: